Learn More
Medchemexpress LLC (3R,4S)-tofacitinib | 1092578-46-5 | MFCD23160057 | 99.9% | 312.37 | C16H20N6O | 10 MG
SDP
Supplier:  Medchemexpress LLC HY40354D10MG
Encompass_Preferred
(3R,4S)-tofacitinib is the less-active enantiomer of the Janus kinase (JAK) inhibitor tofacitinib, supplied as a 10 mg research sample for in vitro and preclinical studies. The compound is provided with supporting analytical documentation (COA, HNMR, LC-MS) and is for research use only, not for human therapeutic use.
  • Enantiomer of tofacitinib with reduced JAK inhibitory activity.
  • High analytical purity (≈99.9%) appropriate for reference and assay work.
  • Documented analytical data: certificate of analysis, NMR, and LC-MS available.
  • Compact 10 mg size convenient for pilot studies and method development.
  • Stable as a crystalline powder; follow supplier handling and storage recommendations.
Catalog No. 50-002-99222