Learn More
Medchemexpress LLC (3R,5R)-3,5-Diacetoxy-1-(3,4-dihydroxyphenyl)-7-(4-hydroxyphenyl)heptane | 1269839-26-0 | ≥95.0% | 416.46 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN90935MG
Encompass_Preferred
(3R,5R)-3,5-Diacetoxy-1-(3,4-dihydroxyphenyl)-7-(4-hydroxyphenyl)heptane is a diarylheptanoid derived from the rhizomes of *Curcuma kwangsiensis*, exhibiting various biological and pharmacological activities.
  • Antioxidant activity
  • Antihepatotoxic activity
  • Anti-inflammatory activity
  • Antiproliferative activity
  • Antiemetic activity
  • Antitumor activities
Catalog No. 50-004-25567