Learn More
Medchemexpress LLC (3R,5S)-5-([1,1′-Biphenyl]-4-ylmethyl)-3-methyl-2-pyrrolidinone | 1038924-70-7 | 95.5% | C18H19NO | 100 MG

Supplier: Medchemexpress LLC HY79080100MG
This biological molecule, (3R,5S)-5-([1,1′-Biphenyl]-4-ylmethyl)-3-methyl-2-pyrrolidinone, is a white to light yellow solid intended for research use only.
- Purity: 95.5%
- Molecular weight: 265.35 g/mol
- Chemical formula: C18H19NO
- Appearance: Solid, white to light yellow
- Solubility in DMSO: 100 mg/mL (376.86 mM)
- Storage as powder: -20°C for 3 years, 4°C for 2 years
- Storage in solvent: -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.