Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (3R,5S)-5-([1,1′-Biphenyl]-4-ylmethyl)-3-methyl-2-pyrrolidinone | 1038924-70-7 | 95.51% | C18H19NO | 50 MG
SDP

Supplier:  Medchemexpress LLC HY7908050MG

Encompass_Preferred

(3R,5S)-5-([1,1′-Biphenyl]-4-ylmethyl)-3-methyl-2-pyrrolidinone is a biological molecule. It is a solid with a white to light yellow color and a molecular weight of 265.35. This product is for research use only.

  • Biological molecule
  • Solid appearance
  • White to light yellow color
  • Purity: 95.51%
  • For research use only
  • Shipped at room temperature in continental US
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-34873


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.