Learn More
Medchemexpress LLC (3R,5S)-5-([1,1'-Biphenyl]-4-ylmethyl)-3-methyl-2-pyrrolidinone | 1038924-70-7 | C18H19NO | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY79080
Encompass_Preferred
(3R,5S)-5-([1,1'-Biphenyl]-4-ylmethyl)-3-methyl-2-pyrrolidinone is a biological molecule intended for research use only. This product has not been fully validated for medical applications.
  • Powder can be stored at -20°C for 3 years or 4°C for 2 years.
  • In solvent, it can be stored at -80°C for 6 months or -20°C for 1 month.
Catalog No. 50-003-59254