Learn More
Medchemexpress LLC (+)-(3R,8S)-Falcarindiol | 225110-25-8 | 260.37 | 25 MG
Supplier: Medchemexpress LLC 25MGHYN1976
(+)-(3R,8S)-Falcarindiol is a polyacetylene found in carrots. It exhibits antimycobacterial activity with an IC50 of 6 μM and MIC of 24 μM against Mycobacterium tuberculosis H37Ra. Additionally, it possesses antineoplastic and anti-inflammatory activities.
- Functions as a click chemistry reagent due to its alkyne group.
- Can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups.
- Suitable for laboratory centrifugation and storage.