Learn More
Medchemexpress LLC (+)-(3R,8S)-Falcarindiol | 225110-25-8 | 260.37 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN19765MG
Encompass_Preferred
(+)-(3R,8S)-Falcarindiol is a polyacetylene naturally found in carrots. This compound exhibits antimycobacterial activity against Mycobacterium tuberculosis H37Ra, with an IC50 of 6 μM and MIC of 24 μM. It also possesses antineoplastic and anti-inflammatory properties, making it a versatile compound for various research applications.
  • Polyacetylene found in carrots.
  • Exhibits antimycobacterial activity.
  • Active against Mycobacterium tuberculosis H37Ra.
  • Possesses antineoplastic activity.
  • Demonstrates anti-inflammatory activity.
  • Functions as a click chemistry reagent.
  • Contains an alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc).
Catalog No. 50-004-16037