Learn More
Medchemexpress LLC (+)-(3R,8S)-Falcarindiol | 225110-25-8 | 260.37 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN197650MG
Encompass_Preferred
(+)-(3R,8S)-Falcarindiol is a polyacetylene derived from carrots, known for its diverse biological activities. It demonstrates potent antimycobacterial effects, alongside antineoplastic and anti-inflammatory properties.
  • Exhibits antimycobacterial activity against Mycobacterium tuberculosis H37Ra.
  • Possesses antineoplastic activity.
  • Functions as an anti-inflammatory agent.
  • Acts as a click chemistry reagent, containing an alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc).
Catalog No. 50-004-15922