Learn More
Medchemexpress LLC (3S)-3-[[(2S)-1-[(2S)-2-[(4-amino-3-chlorobenzoyl)amino]-3,3-dimethylbutanoyl]pyrrolidine-2-car... | 1230628-71-3 | MFCD28386195 | 98.4% | 477.94 g/mol | C22H28ClN5O5 | 50MG

Supplier: Medchemexpress LLC HY1241250MG
ML132 (NCGC-00183434) is a selective caspase-1 inhibitor used as a molecular probe in research on inflammation and cancer. It exhibits an IC50 of approximately 34.9 nM and is provided as a high-purity small-molecule solid for laboratory research applications.
- Selective caspase-1 inhibitor, IC50 ≈ 34.9 nM.
- High reported purity (~98.4%).
- Molecular formula C22H28ClN5O5; molecular weight 477.94 g/mol.
- Stable small molecule suitable as a research probe.
- Used in anti-inflammatory and anticancer pathway studies.
- Store at -80°C, protect from light, under inert atmosphere.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.