Learn More
Medchemexpress LLC [(3S,4R)-3-amino-4-hydroxypiperidin-1-yl]-[2-[1-(cyclopropylmethyl)indol-2-yl]-7-methoxy-1-methylben | 1652629-23-6 | MFCD32688762 | 99.1% | 473.57 g/mol | C27H31N5O3 | 100MG
SDP
Supplier:  Medchemexpress LLC HY123606100MG
Encompass_Preferred
GSK484 is a selective, reversible inhibitor of protein-arginine deiminase 4 (PAD4) used as a chemical probe in inflammation and rheumatoid arthritis research. It reduces inflammation and joint destruction in collagen-induced arthritis models and modulates TNF-α expression, and is supplied as a high-purity solid for biochemical and cellular studies.
  • Selective, reversible PAD4 inhibitor suitable for biochemical assays.
  • Demonstrated activity in arthritis and inflammation models.
  • High purity (99.1%) for research applications.
  • Molecular weight 473.57 g/mol; formula C27H31N5O3.
  • Supplied as a 100 mg solid pack for laboratory use.
Catalog No. 50-002-39471