Learn More
Medchemexpress LLC (3S,4S)-A2-32-01 | 98.4% | 301.42 | 10 MG

Supplier: Medchemexpress LLC HY111532B10MG
(3S,4S)-A2-32-01 (compound 24(S,S)) is a less active (S,S)-enantiomer of A2-32-01. A2-32-01 is a potent caseinolytic protein proteases (ClpP) inhibitor, intended for research use only.
- Less active (S,S)-enantiomer
- Potent ClpP inhibitor
- Oil appearance, colorless to light yellow
- Molecular weight of 301.42
- Chemical formula C19H27NO2
- Recommended storage at -20°C, protected from light
- Soluble in DMSO at 100 mg/mL
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.