Learn More
Medchemexpress LLC (3S,4S)-PF-06459988 | 1858291-14-1 | 98.97% | 431.88 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY19985A
Encompass_Preferred
(3S,4S)-PF-06459988 is the S enantiomer of PF-06459988, exhibiting less activity. It acts as a potent, irreversible inhibitor of T790M mutant epidermal growth factor receptor (EGFR). This compound demonstrates excellent selectivity against EGFR wild-type, featuring a minimally reactive electrophile to reduce off-target labeling.
  • Acts as an S enantiomer of PF-06459988
  • Potent irreversible inhibitor of T790M mutant EGFR
  • Excellent selectivity against EGFR wild-type
  • Minimally reactive electrophile reduces off-target labeling
Catalog No. 50-004-02306