Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (3S,4S)-tivantinib | 905854-03-7 | 98.7% | 369.42 g·mol⁻¹ | C23H19N3O2 | 10 MG
SDP

Supplier:  Medchemexpress LLC HY5068710MG

Encompass_Preferred

(3S,4S)-Tivantinib is the stereochemically defined small-molecule inhibitor of the c-MET receptor tyrosine kinase supplied for laboratory research use. Provided in milligram-scale quantities, it is characterized by standard analytical data and should be stored under recommended conditions to preserve stability.

  • Stereochemically defined (3S,4S) isomer
  • Selective c-MET tyrosine kinase inhibitor for research use
  • Molecular formula C23H19N3O2; molecular weight 369.42 g·mol⁻¹
  • CAS number 905854-03-7 for unambiguous identification
  • Supplied in milligram-scale quantities (common pack: 10 MG)
  • Storage: powder -20°C for long term; 4°C for shorter-term storage

Catalog No. 50-002-98674


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.