Learn More
Medchemexpress LLC (3S,4S)-Tivantinib | 905854-03-7 | 98.71% | C23H19N3O2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY5068750MG
Encompass_Preferred
(3S,4S)-Tivantinib is a potent and highly selective inhibitor of the receptor tyrosine kinase c-MET. It also targets GSK3α and GSK3β, which are important in the cellular mechanism of non-small cell lung cancer (NSCLC).
  • Potent and highly selective inhibitor of c-MET.
  • Targets GSK3α and GSK3β.
  • Important in the cellular mechanism of non-small cell lung cancer (NSCLC).
Catalog No. 50-003-57955