Learn More
Medchemexpress LLC (3S,4S)-Tivantinib | 905854-03-7 | MFCD11977597 | C23H19N3O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY506875MG
Encompass_Preferred
(3S,4S)-Tivantinib is a potent and highly selective inhibitor of the receptor tyrosine kinase c-MET. It also targets GSK3α and GSK3β, which are important in the cellular mechanism of non-small cell lung cancer (NSCLC).
  • Potent and selective c-MET inhibitor
  • Targets GSK3α and GSK3β
  • Plays a role in non-small cell lung cancer (NSCLC) cellular mechanism
Catalog No. 50-003-57743