Learn More
Medchemexpress LLC (3S,4S)-Tofacitinib | 1092578-47-6 | 99.75% | 312.37 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY40354C100MG
Encompass_Preferred
(3S,4S)-Tofacitinib is the less active S-enantiomer of Tofacitinib, which is an inhibitor of JAK3 with an IC50 of 1 nM. This product is intended for research use only.
  • Less active S-enantiomer of Tofacitinib
  • Inhibits JAK3 with an IC50 of 1 nM (for Tofacitinib)
  • Available in solid and solution forms
  • High purity: 99.75%
Catalog No. 50-004-04678