Learn More
Medchemexpress LLC (3S,5R)-Fluvastatin-d6 sodium | 2249799-35-5 | 98.0% | 439.48 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY14664CS1MG
Encompass_Preferred
(3S,5R)-Fluvastatin-d6 (sodium) is the deuterium labeled (3S,5R)-Fluvastatin sodium. Fluvastatin is a first fully synthetic, competitive HMG-CoA reductase inhibitor with an IC50 of 8 nM. Fluvastatin protects vascular smooth muscle cells against oxidative stress through the Nrf2-dependent antioxidant pathway.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-62972