Learn More
Medchemexpress LLC (3S,5R)-fluvastatin sodium | 94061-81-1 | MFCD00929076 | 98.2% | C24H25FNNaO4 | 10MG
SDP
Supplier:  Medchemexpress LLC HY14664C10MG
Encompass_Preferred
(3S,5R)-Fluvastatin sodium is the stereopure sodium salt of the statin fluvastatin supplied for research use as an HMG-CoA reductase inhibitor. It is provided as a high-purity, enantiomerically enriched material intended for biochemical and cellular studies investigating cholesterol biosynthesis, lipid metabolism, and related signaling pathways.
  • High purity and >98% enantiomeric excess.
  • Sodium salt form enhances solubility in assay buffers.
  • Suitable for in vitro enzymatic and cell-based assays.
  • Supplied in small research pack sizes for assay development.
  • Certificate of analysis and safety data sheet available.
Catalog No. 50-002-59557