Learn More
Medchemexpress LLC (3S,5S)-Atorvastatin | 501121-34-2 | 95.0% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB0589C10MM/1ML
Encompass_Preferred
(3S,5S)-Atorvastatin is an inactive enantiomer of Atorvastatin. It can activate pregnane X receptor (PXR). Atorvastatin is an orally active HMG-CoA reductase inhibitor, which effectively decreases blood lipids. This product is for research use only and not sold to patients.
  • Increases CYP2B and CYP3A mRNA content with equal ability as Atorvastatin.
  • Induces luciferase activity with an EC50 of 12.4 μM at 100 μM concentrations.
  • Addition to FPBA/l-tryptophanol mixture induces intensity enhancement of l-tryptophanol fluorescence.
Catalog No. 50-004-07765