Learn More
Medchemexpress LLC (3S,5S)-Atorvastatin | 501121-34-2 | 95.0% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB0589C50MG
Encompass_Preferred
(3S,5S)-Atorvastatin is an inactive enantiomer of Atorvastatin that activates pregnane X receptor (PXR). Atorvastatin is an orally active HMG-CoA reductase inhibitor, known for its ability to effectively decrease blood lipids.
  • Increases CYP2B and CYP3A mRNA content with equal ability as Atorvastatin.
  • Induces luciferase activity with an EC50 of 12.4 μM at 100 μM concentrations.
  • Addition to the 2-formylphenylboronic acid (FPBA)/l-tryptophanol mixture enhances l-tryptophanol fluorescence.
Catalog No. 50-004-08531