Learn More
Medchemexpress LLC (3S,5S)-Pitavastatin calcium | 254452-92-1 | 99.9% | C25H24FNO4·1/2Ca | 10MG
SDP
Supplier:  Medchemexpress LLC HY13538310MG
Encompass_Preferred
(3S,5S)-Pitavastatin calcium is the 3-epimer of pitavastatin calcium, a potent HMG-CoA reductase inhibitor used for research in lipid metabolism and cardiovascular disease. Supplied as a white to off-white solid with high purity and characterized solubility and storage profiles suitable for preclinical and biochemical studies.
  • Potent HMG-CoA reductase inhibitor for biochemical assays.
  • High purity: 99.9%.
  • White to off-white solid appearance for easy handling.
  • Good solubility in DMSO (≈83.3 mg/mL) with ultrasonic aid.
  • Stable when stored sealed at 4°C; solution stability at -80°C for up to 6 months.
Catalog No. 50-002-59809