Learn More
Perkin Elmer US LLC 3-Slot HybridXLT Organics Quartz Torch
Supplier:  Perkin Elmer US LLC N0790249
Encompass_Preferred
PerkinElmer's HybridXLT torch is the latest addition to the family of PerkinElmer genuine sample introduction components. The new hybrid design provides the highest performance for your Avio 200 or 500 spectrometer. This new torch has an extended lifetime of up to 10x your current quartz torch, increasing instrument uptime and decreasing maintenance costs. Features/Benefits: Extended Torch Lifetime - up to 10x improvement. Viewable plasma compared to a full ceramic design. Ideal for all sample types, except organic solvents. Increased instrument uptime. Lower cost per analysis. Lower cost than full ceramic torches.
Catalog No. 50-211-2415