Learn More
BOSTON CELLRON 3T3-msCD40L Mouse Fibroblast Cell Lines
Supplier:  BOSTON CELLRON C1002
Encompass
3T3-CD40L is a mouse fibroblast cell line that secretes mouse CD40L. 3T3-CD40L cells can be co-cultured with B cells to aid B cell expansion.
Catalog No. NC1632519