Learn More
Sigma Aldrich Fine Chemicals Biosciences 3X FLAG Peptide, MFCD01863911, 4mg
Supplier:  Sigma Aldrich Fine Chemicals Biosciences F47994MG
Encompass
Empirical Formula (Hill Notation) C120H169N31O49S, Molecular Weight 2861.87, The 3X FLAG Peptide is a synthetic peptide of 23 amino acid residue. The Asp-Tyr-Lys-Xaa-Xaa-Asp motif is repeated three times in the peptide. Eight amino acids at the C-terminus make up the classic FLAG sequence (Asp-Tyr-Lys-Asp-Asp-Asp-Asp-Lys).
Catalog No. NC1689980