Learn More
Chemglass Life Sciences Chemglass 4.0 mL Cryogenic Vial, Self-Standing, External Thread, Sterile
SDP
Supplier:  Chemglass Life Sciences CGN4762005
Encompass_Preferred
This 4.0 mL cryogenic vial is a sterile, self-standing container with an external thread designed for secure low-temperature storage of biological samples and reagents. It is supplied in racks for organized storage and handling.
  • Holds 4.0 mL.
  • External thread for secure closure.
  • Self-standing design for stability during storage.
  • Sterile, suitable for biological samples.
  • Supplied in racks: 9 x 9 per rack; 8 racks per case; 648 vials per case.
Catalog No. 50-273-3390