Learn More
Roboz Surgical 4-0 SILK BLACK BRAIDED SUTURES
SDP
Supplier:  Roboz Surgical SUT106631
Encompass
Suture Material With C-22 Needle,Sterile, 18 in. Length, Box Of 36. Size: 4-0; Silk, Black, Braided, Non-Absorbable
Catalog No. NC9839314