Learn More
Chemglass Life Sciences Chemglass Cryogenic Vial, Self-Standing, Internal Thread, Sterile, Racked, 4.0 mL (CGN-4766-013)
SDP
Supplier:  Chemglass Life Sciences CGN4766013
Encompass_Preferred
The Chemglass 4.0 mL cryogenic vial (CGN-4766-013) is a sterile, self-standing container designed for cryogenic sample storage and long-term biobanking. Manufactured from USP-6 grade raw materials and sterilized by E-beam in a controlled clean-room environment, the vial features an internal thread closure, clear graduations, a white marking area, and solid one-piece construction. Supplied racked and nested for easy handling, it is intended for storage in the gas phase of liquid nitrogen.
  • Sterile; E-beam sterilized, non-pyrogenic, and DNase/RNase free.
  • 4.0 mL volume with clear graduations and a white marking area.
  • Self-standing round bottom design for upright storage without adapters.
  • Internal thread closure with solid, one-piece construction to minimize barcode loss at low temperatures.
  • Supplied racked and nested (9 x 9) for efficient handling; 8 racks (648 vials) per case.
Catalog No. 50-273-3340