Learn More
Medchemexpress LLC 4-(1-Aminocyclopropyl)benzonitrile hydrochloride | 1369512-65-1 | C10H11ClN2 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY75290500MG
Encompass_Preferred
4-(1-Aminocyclopropyl)benzonitrile hydrochloride is a chemical compound commonly used as a laboratory chemical and in the manufacturing of various substances. This product appears as a solid and is stable under recommended storage conditions, making it suitable for a range of applications.
  • Solid appearance
  • Stable under recommended storage conditions
  • Used as a laboratory chemical
  • Used in the manufacture of substances
  • Recommended powder storage: -20°C for 3 years or 4°C for 2 years
  • Recommended in-solvent storage: -80°C for 6 months or -20°C for 1 month
Catalog No. 50-004-33934