Learn More
Medchemexpress LLC 4-(1-phenylethyl)resorcinol | 85-27-8 | 99.4% | 214.26 g·mol⁻1 | C14H14O2 | 5G
SDP
Supplier:  Medchemexpress LLC HY1258095G
Encompass_Preferred
4-(1-Phenylethyl)resorcinol is a research-grade phenolic compound used as a potent skin-lightening active and biochemical reagent. It inhibits melanin synthesis and is used in formulation and analytical research to evaluate hyperpigmentation treatments and cosmetic efficacy. Supplied as a high-purity solid suitable for weighing, HPLC analysis, and formulation work.
  • High purity suitable for research applications.
  • Proven inhibition of melanin production for skin-lightening studies.
  • Solid powder form for easy weighing and formulation.
  • Available in multiple packaging sizes for small to larger scale studies.
  • Accompanied by datasheet, certificate of analysis, and safety data sheet for quality and safety information.
Catalog No. 50-002-35990