Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-(1-phenylethyl)resorcinol | 85-27-8 | MFCD21648466 | 99.4% | 214.26 g/mol | C14H14O2 | 10G
SDP

Supplier:  Medchemexpress LLC HY12580910G

Encompass_Preferred

4-(1-Phenylethyl)resorcinol is a resorcinol derivative used as a skin-lightening agent and tyrosinase inhibitor for cosmetic and research applications. The compound is supplied as an off-white to yellow solid with high purity and requires storage under inert atmosphere; in solution it is best stored at very low temperatures for long-term stability.

  • High purity: 99.4%.
  • Molecular formula: C14H14O2.
  • Molecular weight: 214.26 g/mol.
  • Appearance: off-white to yellow solid.
  • Storage: store at 4°C under nitrogen; in solvent store at -80°C for up to 6 months.
  • Intended for research use only.

Catalog No. 50-002-36457


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.