Learn More
Medchemexpress LLC 4-(1-phenylethyl)resorcinol | 85-27-8 | MFCD21648466 | 99.4% | 214.26 g/mol | C14H14O2 | 50G
SDP
Supplier:  Medchemexpress LLC HY12580950G
Encompass_Preferred
4-(1-Phenylethyl)resorcinol is a small-molecule tyrosinase inhibitor used in cosmetic and pigmentation research to reduce melanin synthesis. Supplied as a solid for research use, it is typically applied in in vitro assays and formulation studies to evaluate skin-lightening activity and mechanism of action.
  • Tyrosinase inhibitor that reduces melanin production.
  • High reported purity (99.4%), suitable for research applications.
  • Solid form, stable under recommended storage conditions.
  • Molecular formula C14H14O2 and molecular weight 214.26 g/mol.
  • Used in pigmentation studies, cosmetic formulation, and biochemical assays.
Catalog No. 50-002-35392