Learn More
Medchemexpress LLC 4 10-Dioxatri cyclo 100g
SDP
Supplier:  Medchemexpress LLC HYW102981100G
Encompass_Preferred
4 10-Dioxatri cyclo[5 2 1 02 6]dec-8-ene-3 5-dione is a purine nucleoside analog Purine nucleoside analogs have broad antitumor activity targeting indolent lymphoid malignancies Anticancer mechanisms in this process rely on inhibition of DNA synthesis induction of apoptosis etc[1]
Catalog No. 50-002-06510