Learn More
Medchemexpress LLC 4-1BB/TNFRSF9 Mouse 10ug
SDP
Supplier:  Medchemexpress LLC HYP7288410UG
Encompass_Preferred
4-1BB (CD137 TNFRSF9) a receptor of TNFSF9/4-1BBL belongs to the tumor necrosis factor (TNF) receptor superfamily[1] 4-1BB is helpful for T cell activation and development and also induces peripheral mononuclear cell proliferation and migration to the tumor microenvironment[2] 4-1BB is also involved in enhancing Nrf2 and NF- B pathway mediated apoptosis of endothelial cells[3] Mouse 4-1BB protein is a surface glycoprotein with a transmembrane domain 4-1BB/TNFRSF9 Protein Mouse (HEK293 His) is the extracellular part (V24-L187) of 4-1BB protein produced by HEK293 cells with C-terminal His-tag
Catalog No. 50-001-81831