Learn More
Medchemexpress LLC 4-1BB/TNFRSF9 protein, mouse (C-hFc fusion) | ≥95.0% | 500 UG
SDP
Supplier:  Medchemexpress LLC HYP7447A500UG
Encompass_Preferred
Recombinant mouse 4-1BB (TNFRSF9) Fc-fusion protein produced in HEK293 cells. Supplied lyophilized for research use in biochemical and cell-based assays, it serves as a reagent for receptor-ligand interaction studies and T-cell costimulation research.
  • Produced in HEK293 cells.
  • C-terminal human Fc fusion (C-hFc).
  • Residues V24-L187 (accession P20334).
  • Purity ≥95.0% by reducing SDS-PAGE.
  • Lyophilized from 20 mM MES, 150 mM NaCl, pH 5.5.
  • Reconstitute to ≥100 μg/mL in ddH2O; avoid lower concentrations.
  • Approximate molecular weight ~58.7 kDa; biological activity ED50 ≈0.603 μg/mL.
  • Store lyophilized at -20 °C; freeze aliquots at -20 °C or -80 °C for extended storage.
Catalog No. 50-002-93460