Learn More
Medchemexpress LLC 4-1bbl/tnfsf9 protein, mouse (HEK293, His) | ≥98.0% | 500 UG

Supplier: Medchemexpress LLC HYP7446500UG
Recombinant mouse 4-1BB ligand (TNFSF9) expressed in HEK293 cells with a His tag. Supplied lyophilized for stability, this biologically active protein is intended for immunology and cell-signaling applications involving 4-1BB/CD137 receptor activation and T-cell co-stimulation.
- Expressed in HEK293 mammalian cells.
- Contains His tag for detection and purification.
- Lyophilized formulation for improved stability.
- Reconstitution recommended to 100-200 μg/mL in ddH2O.
- Endotoxin level <1 EU/μg.
- Purity ≥98.0% as determined by certificate of analysis.
- Stable at -20°C for long-term storage; aliquot and avoid repeated freeze-thaw cycles.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.