Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-(1H-pyrazol-4-yl)-7H-pyrrolo[2,3-d]pyrimidine | 952518-97-7 | MFCD20683197 | 97.0% | 185.19 g/mol | C9H7N5 | 25 MG
SDP

Supplier:  Medchemexpress LLC 25MGHYW059210

Encompass_Preferred

Ruxolitinib impurity-1 (CAS 952518-97-7) is a characterized impurity of ruxolitinib used as an analytical reference standard for impurity profiling and safety analysis. It is typically supplied in small vial quantities for laboratory analytical use.

  • Purity: 97.0%.
  • CAS number: 952518-97-7.
  • Molecular formula: C9H7N5.
  • Molecular weight: 185.19 g/mol.
  • Intended use: impurity profiling and safety analysis.
  • Pack size: 25 MG.

Catalog No. 50-002-09080


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.