Learn More
Medchemexpress LLC 4-((1R,2R)-2-hydroxycyclohexyl)-2-(trifluoromethyl)benzonitrile | 1076225-26-7 | 99.7% | 269.26 g/mol | C14H14F3NO | 5 MG
SDP
Supplier:  Medchemexpress LLC HY14250A5MG
Encompass_Preferred
(Rac)-PF-998425 is a potent, selective, nonsteroidal androgen receptor (AR) antagonist provided for research use. It exhibits an AR binding IC50 of 26 nM and a cellular IC50 of 90 nM, and is frequently used in studies of androgenetic alopecia and AR signaling. Chemical properties include CAS 1076225-26-7, molecular formula C14H14F3NO, molecular weight 269.26 g/mol, and typical purity of 99.68%.
  • Potent AR antagonist with nanomolar activity.
  • Selective, nonsteroidal modulation of androgen receptor.
  • High purity suitable for biochemical and cellular assays.
  • Available as solid and as DMSO solution formats for convenience.
  • Relevant for androgenetic alopecia and AR signaling research.
Catalog No. 50-002-21731