Learn More
Medchemexpress LLC 4-[2,2-bis(diethoxyphosphoryl)ethenyl]-2,6-ditert-butylphenol | 126411-39-0 | MFCD00888473 | ≥98.0% | 504.54 g/mol | C24H42O7P2 | 200 MG

Supplier: Medchemexpress LLC HY100793200MG
SR12813 is a small-molecule HMG-CoA reductase inhibitor and a potent agonist of the human pregnane X receptor (hPXR), used in biochemical and pharmacological research. It preferentially binds human PXR over mouse PXR and is supplied for research use only.
- Inhibits HMG-CoA reductase (IC50 = 0.85 μM).
- Acts as an efficient human pregnane X receptor (hPXR) agonist.
- Selective for human PXR over mouse PXR.
- Chemical formula C24H42O7P2 and molecular weight 504.54 g/mol.
- Purity reported as ≥98% by HPLC where specified.
- Intended for research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.