Learn More
Medchemexpress LLC 4-[2-[4-(2-pyridin-2-yl-5,6-dihydro-4H-pyrrolo[1,2-b]pyrazol-3-yl)quinolin-7-yl]oxyethyl]morpholine | 700874-71-1 | ≥98.0% | 50 MG

Supplier: Medchemexpress LLC HY1207550MG
LY2109761 is an orally active small-molecule dual inhibitor of transforming growth factor-β (TGF-β) receptor type I and II used for research applications. It shows Ki values of 38 nM for TβRI and 300 nM for TβRII, and is supplied as a powder for analytical and research use.
- Orally active TGF-β receptor type I/II inhibitor
- Ki values of 38 nM (TβRI) and 300 nM (TβRII)
- Powder form suitable for analytical and research use
- Molecular formula C26H27N5O2; molecular weight 441.52 g/mol
- Soluble in DMSO (6.67 mg/mL) and suspendable in 0.5% CMC-Na/0.5% Tween-80 (1 mg/mL)
- Stable when stored as a powder at -20°C for long-term storage
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.