Learn More
Medchemexpress LLC 4-(((2,4-diaminopteridin-6-yl)methyl)(methyl)amino)benzoic acid | 19741-14-1 | MFCD00075774 | 98.4% | 325.33 g/mol | C15H15N7O2 | 25 MG

Supplier: Medchemexpress LLC HY10825125MG
Methotrexate metabolite (DAMPA) is an analytical reference standard representing the primary metabolite of methotrexate, provided as a solid for research and impurity-profiling applications. It is intended for use in method development and quantitative/qualitative analysis by chromatographic and spectrometric techniques.
- Analytical standard for LC, GC, and mass spectrometry.
- Purity 98.4% (as listed).
- Molecular weight 325.33 g/mol.
- Chemical formula C15H15N7O2.
- Appearance solid, yellow to brown.
- Typical package size 25 MG.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.