Learn More
Medchemexpress LLC 4-(2,4-Difluorophenoxy)benzoic acid | 748183-47-3 | 98.95% | 250.20 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY43859
Encompass_Preferred
This chemical is 4-(2,4-Difluorophenoxy)benzoic acid, a white to off-white solid primarily used as a laboratory chemical and in the manufacture of various substances. It is stable under recommended storage conditions and requires specific temperatures for long-term storage.
  • Identified for laboratory use
  • Suitable for manufacturing processes
  • Ships at room temperature for durations less than two weeks
  • Powder stable for 3 years at -20°C or 2 years at 4°C
  • In solvent, stable for 6 months at -80°C or 1 month at -20°C
Catalog No. 50-004-03284