Learn More
Medchemexpress LLC 4-(2-methyl-3-propan-2-ylimidazol-4-yl)-N-(4-methylsulfonylphenyl)pyrimidin-2-amine | 602306-29-6 | MFCD11112135 | 99.9% | 371.46 g/mol | C18H21N5O2S | 500 MG
SDP
Supplier:  Medchemexpress LLC HY10012500MG
Encompass_Preferred
AZD-5438 is a small-molecule inhibitor of cyclin-dependent kinases CDK1, CDK2, and CDK9, characterized by low nanomolar potency in biochemical assays. It is used in research on cell-cycle regulation, cancer pharmacology, and radiosensitization studies, and has a molecular formula of C18H21N5O2S with a molecular weight of 371.46 g/mol.
  • Potent inhibition of CDK1, CDK2, and CDK9 with reported IC50s of 16 nM, 6 nM, and 20 nM.
  • High purity (>99.9% by HPLC) suitable for biochemical and cell-based assays.
  • Supplied as a 500 mg research-grade solid for laboratory use.
  • Molecular formula C18H21N5O2S and molecular weight 371.46 g/mol for precise dosing and calculations.
  • Applicable for studies of cell-cycle regulation, cancer mechanisms, and radiosensitization.
Catalog No. 50-002-15177