Learn More
Accela Chembio Inc 4-(2-nitrophenyl)thiazole-2-amine | 1g | 90323-06-1 | MFCD02352318 | 97+% | Shelf Life: 2070 Days | Light Sensitive
Supplier:  Accela Chembio Inc SY0049801G
Encompass_Preferred
4-(2-nitrophenyl)thiazole-2-amine | 1g | 90323-06-1 | MFCD02352318 | 97+% | Shelf Life: 2070 Days | Light Sensitive
Catalog No. 50-250-639