Learn More
Medchemexpress LLC 4-((21S,24S)-1-(bicyclo[6.1.0]non-4-yn-9-yl)-21-isopropyl-3,19,22-trioxo-24-(3-ureidopropyl)) | 2762519-08-2 | MFCD32062688 | 98.8% | 1546.93 g·mol⁻¹ | C80H127N11O19 | 1 MG

Supplier: Medchemexpress LLC HY1411551MG
endo-BCN-PEG4-Val-Cit-PAB-MMAE is a cleavable antibody-drug conjugate (ADC) linker containing an endo-bicyclononyne (BCN) click handle and a PEG4 spacer conjugated to monomethyl auristatin E (MMAE). It is used for ADC synthesis and for bioconjugation via strain-promoted alkyne-azide cycloaddition (SPAAC).
- Cleavable Val-Cit-PAB linker for enzymatic release of payload.
- Contains a PEG4 spacer to improve solubility and linker flexibility.
- Endo-BCN group enables copper-free click conjugation to azide-bearing molecules.
- Conjugated to MMAE for use in antibody-drug conjugate research.
- Supplied in small milligram quantities for research and synthesis workflows.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.