Learn More
Medchemexpress LLC 4-[[(2R)-1-(1-benzothiophene-3-carbonyl)-2-methylazetidine-2-carbonyl]-[(3-chlorophenyl)methyl]amino]but | 1391076-61-1 | MFCD28502087 | 98.1% | 485.00 | C25H25ClN2O4S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY129405MG
Encompass_Preferred
GLPG0974 is a small-molecule antagonist of the free fatty acid receptor 2 (FFA2/GPR43) provided as a research reagent. It shows potent activity in receptor assays (IC50 9 nM) and is supplied as a high-purity solid for in vitro pharmacology and biochemical studies. For research use only; not intended for human use.
  • Potent FFA2/GPR43 antagonism with IC50 9 nM.
  • High purity suitable for biochemical and cell-based assays.
  • Supplied as a solid for convenient handling and storage.
  • Characterized compound with CAS 1391076-61-1 and molecular weight 485.00.
  • Intended for laboratory research applications only.
Catalog No. 50-002-62902