Learn More
Medchemexpress LLC 4-({[3-(4-carboxybenzyl)-1-methyl-2-oxo-1,2,3,4-tetrahydroquinolin-6-yl]methyl}amino)benzoic acid | 1222173-37-6 | 99.1% | 446.46 g/mol | C24H22N4O5 | 10MM 1ML
SDP
Supplier:  Medchemexpress LLC HY12402910MM 1ML
Encompass_Preferred
BI-4394 is a potent, selective, and orally active inhibitor of matrix metalloproteinase-13 (MMP-13) used as a chemical probe in research on MMP-related biology. It is supplied as a 10 mM solution in DMSO and is characterized by high purity and defined solubility and storage recommendations.
  • Potent MMP-13 inhibition (IC50 1 nM).
  • Greater than 1000-fold selectivity over other MMPs.
  • Supplied as a 10 mM solution in DMSO, 1 mL.
  • High purity (99.09%).
  • Soluble in DMSO up to 100 mg/mL (≈223.98 mM) with warming.
  • Stable under recommended storage conditions for powder and solution.
  • Includes IC50 data for related MMPs (MMP-14, MMP-9, MMP-10, MMP-2).
Catalog No. 50-002-38411