Learn More
Sklar Instruments Single Use, Econo-Sterile, Adson Dressing Forceps, Sklar 4-3/4 in., Blunt, Straight, Serrated, Stainless Steel, Disposable, CS of 25.
SDP
Supplier:  Sklar Instruments 962571
Encompass_Preferred
Single Use, Econo-Sterile, Adson Dressing Forceps, Sklar, 4-3/4 in., Blunt, Straight, Serrated, Stainless Steel, Disposable, CS of 25. Adson Dressing Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds.
Catalog No. 50-109-4283