Learn More
Sklar Instruments Scissors, Sklar Plastic Surgery Scissors, Sklar 4-3/4 in., Straight, Razor, Stainless Steel, Reusable, EA.
SDP
Supplier:  Sklar Instruments 153460
Encompass_Preferred
Scissors, Sklarhone Plastic Surgery Scissors, Sklar, 4-3/4 in., Straight, Razor, Stainless Steel, Reusable, EA. Sklarhone Plastic Surgery Scissors are non-ratcheted, finger ring scissors used for cutting and dissecting delicate tisssue in various plastic surgery procedures. Sklarhone scissors feature one knife-like razor sharp edge and one standard scissor edge. This allows the user to make the sharp incisions of a knife with the cutting ability of scissors. Catching skin and pulls are minimized by the use of Sklarhone premium scissors. These instruments are identified by dual black finger rings. This product is straight with smooth blades, sharp/sharp tips and an overall length of 4-3/4 inches.
Catalog No. 50-109-3452