Learn More
Medchemexpress LLC 4-(3-(5-chloro-2-methoxyphenyl)thioxureido)-1-ethyl-1H-pyrazole-3-carboxamide | 957485-64-2 | 99.3% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY12281100MG
Encompass_Preferred
FPH2 is a small-molecule research compound that induces functional proliferation of primary human hepatocytes and is used in studies toward therapeutics for liver diseases. In vitro, it increases hepatocyte nuclei count and the number of nuclei undergoing mitosis in a concentration-dependent manner while maintaining liver-specific functions.
  • Induces proliferation of primary human hepatocytes.
  • Maintains liver-specific functions during proliferation.
  • Shows concentration-dependent activity in vitro.
  • High purity suitable for research applications.
  • Stable as a powder when stored at -20°C for long-term storage.
Catalog No. 50-002-31341