Learn More
Medchemexpress LLC 4-((3,5-dichloro-2-hydroxybenzyl)amino)-2-hydroxybenzoic acid | 1181226-02-7 | 99.2% | 328.15 g/mol | C14H11Cl2NO4 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10045625MG
Encompass_Preferred
ZL006 is a small-molecule research inhibitor that disrupts the interaction between neuronal nitric oxide synthase (nNOS) and postsynaptic density protein-95 (PSD-95), reducing NMDA receptor-mediated nitric oxide (NO) production. It is used in preclinical studies to evaluate neuroprotection and modulation of excitotoxic signaling.
  • Selective disruption of nNOS-PSD-95 interaction.
  • Reduces NMDA receptor-mediated nitric oxide synthesis.
  • Demonstrated neuroprotective activity in preclinical models.
  • High chemical purity suitable for research applications.
  • Suitable for in vitro and in vivo preclinical studies.
Catalog No. 50-002-14082